Learn More
Supertek™ Music Tuning Fork Set of 8 with Activator
GREENER_CHOICESCI_ED
Catalog No. S81836 Shop All Supertek Scientific Products
Change view
Click to view available options
Includes:
8 Tuning Forks, Activator and Case
Catalog No. Includes
S81836 8 Tuning Forks, Activator and Case
1 options
This item is not returnable. View return policy
Catalog No. S81836 Supplier Supertek™ Supplier No. PH36128
Add to Cart
Add to Cart
This item is not returnable. View return policy
Supertek™ Music Tuning Fork Set of 8 with Activator is Superior Quality. The set is tuned to correspond to a 'Tempered' C Major Scale. This kit comes in a zippered canvas carrying case and also includes a rubber activator, to avoid damaging the forks.
  • Includes 1 each of:
    • 261.6 Hz,'C'
    • 293.7 Hz, 'D'
    • 329.6 Hz, 'E'
    • 349.2 Hz, 'F'
    • 392 Hz, 'G'
    • 440 Hz, 'A'
    • 493.9 Hz, 'B'
    • 523.2 Hz, 'C'
    • Zippered canvas carrying case
    • Rubber activator
Specifications
Product Type Tuning Fork
Material Aluminum
Includes 8 Tuning Forks, Activator and Case
Dimensions (L x W x H) 27.9 x 17.5 x 4.6 cm
Weight (Metric) 840 g
Frequency 261.6, 293.7, 329.6, 349.2, 392, 440, 493.9, 523.2 Hz