Learn More
Supertek™ Musical Superior Grade 341.3 Hz 'F' Tuning Fork
GREENER_CHOICESCI_ED
Catalog No. S81854 Shop All Supertek Scientific Products
Change view
Click to view available options
Frequency:
341.3 Hz
Catalog No. Frequency
S81854 341.3 Hz
1 options
This item is not returnable. View return policy
Catalog No. S81854 Supplier Supertek™ Supplier No. PH36121/6M
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Supertek™ Musical Superior Grade 341.3 Hz 'F' Tuning Fork is non-magnetic, corrosion-resistant aluminum alloy construction for light weight and durability. Superior performance and consistent frequency accuracy. Delivers clear tone, longer sound and accurate tuning.
  • Tuning fork is stamped with frequency and tone
Specifications
Product Type Tuning Fork
Material Aluminum
Frequency 341.3 Hz