Learn More
Supertek™ Musical Superior Grade 493.9 Hz 'B' Tuning Fork
GREENER_CHOICESCI_ED
Catalog No. S81858 Shop All Supertek Scientific Products
Change view
Click to view available options
Frequency:
493.9 Hz
Catalog No. Frequency
S81858 493.9 Hz
1 options
This item is not returnable. View return policy
Catalog No. S81858 Supplier Supertek™ Supplier No. PH36121/9M
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Supertek™ Musical Superior Grade 493.9 Hz 'B' Tuning Fork is non-magnetic, corrosion-resistant aluminum alloy construction for light weight and durability. Superior performance and consistent frequency accuracy. Delivers clear tone, longer sound and accurate tuning.
  • Tuning fork is stamped with frequency and tone
Specifications
Product Type Tuning Fork
Material Aluminum
Frequency 493.9 Hz