Learn More
Supertek™ Musical Superior Grade 523.3 Hz 'C' Tuning Fork
GREENER_CHOICESCI_ED
Catalog No. S81859 Shop All Supertek Scientific Products
Change view
Click to view available options
Frequency:
523.3 Hz
Catalog No. Frequency
S81859 523.3 Hz
1 options
This item is not returnable. View return policy
Catalog No. S81859 Supplier Supertek™ Supplier No. PH36121/10M
Add to Cart
Add to Cart
This item is not returnable. View return policy
This Supertek™ Musical Superior Grade 523.3 Hz 'C' Tuning Fork, one octave above middle 'C', is non-magnetic, corrosion-resistant aluminum alloy construction for light weight and durability. Superior performance and consistent frequency accuracy. Delivers clear tone, longer sound and accurate tuning.
  • Tuning fork is stamped with frequency and tone
Specifications
Product Type Tuning Fork
Material Aluminum
Frequency 523.3 Hz