Learn More
Carnabio Usa Inc MUSK/10UG/CATALYTIC
Supplier:  Carnabio Usa Inc 0815310
Encompass
MUSK, Catalytic domain, Wild type, Amino Acid 527-869, NP_005583.1, Expressed in Insect (sf21); 10microg
Catalog No. 50-754-0891