Learn More
Carnabio Usa Inc MUSK/5UG/CATALYTIC
Supplier:  Carnabio Usa Inc 081535
Encompass
MUSK, Catalytic domain, Wild type, Amino Acid 527-869, NP_005583.1, Expressed in Insect (sf21); 5microg
Catalog No. 50-754-0532