Learn More
Medchemexpress LLC Musk ketone | 81-14-1 | 294.31 | 1 ML
SDP
Catalog No. 5000415685
Encompass_Preferred
Musk ketone is an artificial fragrance and a cytochrome P450 enzyme inducer. It exhibits mutagenic and comutagenic effects in Hep G2 cells and promotes neural stem cell proliferation and differentiation in cerebral ischemia by activating the PI3K/Akt signaling pathway. In the brain, musk ketone provides neuroprotection against stroke injury by inhibiting cell apoptosis.
  • Enhances proliferation and differentiation in neural stem cells (NSCs) with increased PI3K/Akt signaling pathway activation.
  • Inhibits cell viability of GC cells and prevents colony formation.
  • Induces cell cycle arrest and apoptosis in AGS and HGC-27 cells.
  • Ameliorates neurological deficit and reduces infarction and cerebral ischemic injury.
  • Increases CYP2B activity, protein, and mRNA levels, and boosts CYP1A enzyme activity.
Catalog No. 50-004-15685 Supplier Medchemexpress LLC Supplier No. HYN204510MM/1ML
Musk ketone is an artificial fragrance and a cytochrome P450 enzyme inducer. It exhibits mutagenic and comutagenic effects in Hep G2 cells and promotes neural stem cell proliferation and differentiation in cerebral ischemia by activating the PI3K/Akt signaling pathway. In the brain, musk ketone provides neuroprotection against stroke injury by inhibiting cell apoptosis.
  • Enhances proliferation and differentiation in neural stem cells (NSCs) with increased PI3K/Akt signaling pathway activation.
  • Inhibits cell viability of GC cells and prevents colony formation.
  • Induces cell cycle arrest and apoptosis in AGS and HGC-27 cells.
  • Ameliorates neurological deficit and reduces infarction and cerebral ischemic injury.
  • Increases CYP2B activity, protein, and mRNA levels, and boosts CYP1A enzyme activity.