Learn More
Medchemexpress LLC Musk ketone | 81-14-1 | 294.31 | 50 MG
SDP
Catalog No. 5000414842
Encompass_Preferred
Musk ketone is an artificial fragrance and a cytochrome P450 enzyme inducer. It demonstrates mutagenic and comutagenic effects in Hep G2 cells and can induce neural stem cell proliferation and differentiation in cerebral ischemia through activation of the PI3K/Akt signaling pathway. It also exhibits neuroprotective properties against stroke injury by inhibiting cell apoptosis.
  • Enhances proliferation and differentiation in neural stem cells.
  • Inhibits cell viability and colony formation in GC cells.
  • Induces cell cycle arrest and apoptosis in AGS and HGC-27 cells.
  • Ameliorates neurological deficit and reduces infarction in rats.
  • Increases CYP2B and CYP1A enzyme activity.
Catalog No. 50-004-14842 Supplier Medchemexpress LLC Supplier No. HYN204550MG
Musk ketone is an artificial fragrance and a cytochrome P450 enzyme inducer. It demonstrates mutagenic and comutagenic effects in Hep G2 cells and can induce neural stem cell proliferation and differentiation in cerebral ischemia through activation of the PI3K/Akt signaling pathway. It also exhibits neuroprotective properties against stroke injury by inhibiting cell apoptosis.
  • Enhances proliferation and differentiation in neural stem cells.
  • Inhibits cell viability and colony formation in GC cells.
  • Induces cell cycle arrest and apoptosis in AGS and HGC-27 cells.
  • Ameliorates neurological deficit and reduces infarction in rats.
  • Increases CYP2B and CYP1A enzyme activity.