Learn More
Medchemexpress LLC MUSK Mouse Myc Hi 10ug | 10ug
SDP
Catalog No. 5000196381
Encompass_Preferred
MUSK protein is a receptor tyrosine kinase that plays a critical role in the formation of the neuromuscular junction (NMJ) MUSK is activated by the AGRIN-LRP4 complex and regulates the NMJ through gene expression actin cytoskeleton reorganization and acetylcholine receptor (AChR) clustering MUSK Protein Mouse (Myc His-SUMO) is the recombinant mouse-derived MUSK protein expressed by E coli with C-Myc N-SUMO N-10 His labeled tag
Catalog No. 50-001-96381 Supplier Medchemexpress LLC Supplier No. HYP7156810UG
MUSK protein is a receptor tyrosine kinase that plays a critical role in the formation of the neuromuscular junction (NMJ) MUSK is activated by the AGRIN-LRP4 complex and regulates the NMJ through gene expression actin cytoskeleton reorganization and acetylcholine receptor (AChR) clustering MUSK Protein Mouse (Myc His-SUMO) is the recombinant mouse-derived MUSK protein expressed by E coli with C-Myc N-SUMO N-10 His labeled tag