Learn More
Medchemexpress LLC MUSK Mouse Myc Hi 50ug
SDP
Catalog No. 5000196442
Encompass_Preferred
Recombinant fragment of mouse Muscle-Specific Kinase (MuSK) with Myc and His-SUMO tags supplied as a lyophilized powder. The protein is expressed in E. coli and formulated for stability, with low endotoxin and manufacturer guidance for reconstitution, storage, and handling available in the product datasheet.
  • Recombinant fragment covering amino acids E22-S494.
  • Myc and His-SUMO affinity tags for detection and purification.
  • Expressed in E. coli for high yield production.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Lyophilized formulation for extended shelf life and easier shipping.
  • Stable at -20°C for up to two years from receipt when stored properly.
Catalog No. 50-001-96442 Supplier Medchemexpress LLC Supplier No. HYP7156850UG
Recombinant fragment of mouse Muscle-Specific Kinase (MuSK) with Myc and His-SUMO tags supplied as a lyophilized powder. The protein is expressed in E. coli and formulated for stability, with low endotoxin and manufacturer guidance for reconstitution, storage, and handling available in the product datasheet.
  • Recombinant fragment covering amino acids E22-S494.
  • Myc and His-SUMO affinity tags for detection and purification.
  • Expressed in E. coli for high yield production.
  • Low endotoxin level (<1 EU/μg) suitable for cell-based assays.
  • Lyophilized formulation for extended shelf life and easier shipping.
  • Stable at -20°C for up to two years from receipt when stored properly.