Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC MUSK Mouse Myc Hi 5ug
SDP

Catalog No. 5000196382
Encompass_Preferred

Recombinant mouse MUSK protein bearing N-terminal Myc and His-SUMO tags, supplied lyophilized for biochemical and biophysical studies. The product is a fragment corresponding to residues E22-S494, expressed in E. coli, and formulated for stability and downstream assays such as SPR after buffer exchange.

  • Recombinant fragment covering residues E22-S494.
  • Expressed in E. coli and supplied lyophilized for long-term storage.
  • Endotoxin level below 1 EU/μg as measured by LAL.
  • Lyophilized from Tris-EDTA with trehalose for enhanced stability.
  • Recommended reconstitution at not less than 100 μg/mL; buffer exchange suggested for SPR.
  • Approximate molecular weight 69.3 kDa.

Catalog No. 50-001-96382 Supplier Medchemexpress LLC Supplier No. HYP715685UG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Recombinant mouse MUSK protein bearing N-terminal Myc and His-SUMO tags, supplied lyophilized for biochemical and biophysical studies. The product is a fragment corresponding to residues E22-S494, expressed in E. coli, and formulated for stability and downstream assays such as SPR after buffer exchange.

  • Recombinant fragment covering residues E22-S494.
  • Expressed in E. coli and supplied lyophilized for long-term storage.
  • Endotoxin level below 1 EU/μg as measured by LAL.
  • Lyophilized from Tris-EDTA with trehalose for enhanced stability.
  • Recommended reconstitution at not less than 100 μg/mL; buffer exchange suggested for SPR.
  • Approximate molecular weight 69.3 kDa.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.