Learn More
MOLECULAR DEPOT LLC Mussel Adhesive Protein - 5 mg
Catalog No. 5000013367
Encompass_Preferred
Mussel Adhesive Protein (MAP) is a highly purified tyrosinase-pretreated Mussel (Mytilus edulis) foot protein-1 This product has been used as coat any surfaces for immobilization of cells and tissue It has also been used in a wide array of other biochemical and immunological applications For in vitro research use only
Catalog No. 50-000-13367 Supplier MOLECULAR DEPOT LLC Supplier No. B2011021
Mussel Adhesive Protein (MAP) is a highly purified tyrosinase-pretreated Mussel (Mytilus edulis) foot protein-1 This product has been used as coat any surfaces for immobilization of cells and tissue It has also been used in a wide array of other biochemical and immunological applications For in vitro research use only