Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MCR Safety Mustang™ Premium Select Side Cow Leather Welding Gloves

Catalog No. 19069569 Shop All MCR Safety Products
Change view
Click to view available options
Coating Material:
Uncoated
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Coating Material
19-069569 Uncoated
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 19-069569 Supplier MCR Safety Supplier No. 4700
This item is not returnable. View return policy

Gloves are sewn with Aramid Fiber to provide maximum protection against heat, flames, and sparks.

  • Premium quality select side cow leather
  • Sewn with Aramid Fiber thread
  • Full 100% cotton sock lining
  • One piece, 13 in. back
  • Fully welted

Specifications

Coating Material Uncoated
Color Gray
Grade Premium
Material Side Split Cowhide Leather
Size X-Large
Temperature (English) Max. 390°F
Temperature (Metric) Max 200°C
For Use With (Application) Welding
Glove Cuff Style Gauntlet
Puncture Rating ANSI/ISEA 105-16 Level 5
Series Mustang™ 4700
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.