Learn More
Medchemexpress LLC HY-18169 5mg Medchemexpress, MUT056399 CAS:1269055-85-7 Purity:>98%
SDP
Catalog No. 502082023
Encompass_Preferred
Medchemexpress, HY-18169 5mg MUT056399 CAS:1269055-85-7 MUT056399 (Fab-001) is a highly potent inhibitor of the FabI enzyme of both S. aureus and E. coli with 50% inhibitory concentration IC50s of 12 nM and 58 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-2023 Supplier Medchemexpress LLC Supplier No. HY181695MG
Medchemexpress, HY-18169 5mg MUT056399 CAS:1269055-85-7 MUT056399 (Fab-001) is a highly potent inhibitor of the FabI enzyme of both S. aureus and E. coli with 50% inhibitory concentration IC50s of 12 nM and 58 nM, respectively. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.