Learn More
Description
mutaFISH™ AR-V7 ARwt RNA Probes is designed to detect AR wildtype and AR-V7 on single strand RNA in cells using padlock probe and in siturolling-circle amplification technology.
Includes:
- RT AR-V7 Primer
- RT ARwt Primer
- mutaFISH™ AR-V7 RNA Probe
- mutaFISH™ ARwt RNA Probe
- Detection Probe-Cy3
- Detection Probe-Cy5
Specifications
Specifications
| Content And Storage | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Detection Method | Fluorescence |
| Format | Kit |
| Label or Dye | Cy3, Cy5 |
| For Use With (Application) | mutaFISH-Ce,mutaFISH-Fr |
| Probe | AR-V7,Arwt |
| Product Type | mutaFISH™ (mutation-specific Fluorescence In Situ Hybridization) |
| Quantity | 1 Probe Set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.