Learn More
Description
mutaFISH™ CEP1wt DNA Probes is designed to identify human CEP1 amplification on dsDNA in cells using padlock probe and in siturolling-circle amplification technology.
- mutaFISH™ CEP1wt DNA Probe
- Detection Probe-FITC
Specifications
Specifications
| Content And Storage | Store at -20°C. Aliquot to avoid repeated freezing and thawing. |
| Detection Method | Fluorescence |
| Format | Kit |
| Label or Dye | FITC |
| For Use With (Application) | mutaFISH-Ce |
| Probe | CEP1wt |
| Product Type | mutaFISH™ (mutation-specific Fluorescence In Situ Hybridization) |
| Quantity | 1 Probe Set |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.