Learn More
Abnova™ mutaFISH™ CK7wt RNA Probes
Catalog No. 89339112 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
mutaFISH-Ce,mutaFISH-P
Catalog No. For Use With (Application)
89-339-112 mutaFISH-Ce,mutaFISH-P
1 options
Catalog No. 89-339-112 Supplier Abnova™ Supplier No. FP0027
Add to Cart
Add to Cart
mutaFISH™ CK7wt RNA Probes is designed to detect human CK7 gene on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.
Specifications
For Use With (Application) mutaFISH-Ce,mutaFISH-P
Includes Content:
1. RT CK7 Primer
2. mutaFISH™ CK7 RNA Probe
3. Detection Probe-FITC