Learn More
Abnova™ mutaFISH™ DNA Accessory Kit 2 for Cells
Catalog No. 89333043 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Kit
Catalog No. Quantity
89-333-043 1 Kit
1 options
Catalog No. 89-333-043 Supplier Abnova™ Supplier No. KA4928
Add to Cart
Add to Cart
mutaFISH™ DNA Accessory Kit 2 for Cells provides reagents and enzymes for in situ restriction digestion, exonucleolysis, mutaFISH™ hybridization, ligation and amplification in cell prior to mutaFISH™.
Box 1 content:
1. H2O
2. PBS
3. Wash Buffer 1
4. Digestion Enzyme Buffer
5. Wash Buffer 2
6. Formamide
7. Detection Buffer
Box 2 content:
1. Digestion Enzyme
2. BSA
3. Restriction Enzyme 2
4. 10X Restriction Enzyme Buffer
5. Hybrid Enzyme
6. 10X Hybrid Enzyme Buffer
7. 1M KCL
8. DNA Polymerase
9. 10X DNA Pol Buffer
10. dNTP Mix
11. 50% Glycerol
12. DAPI (100uM)
Specifications
Content And Storage Store Box 1 at 4°C, store Box 2 at -20°C.
Quantity 1 Kit