Learn More
Abnova™ mutaFISH™ EGFRwt RNA Probes
Catalog No. 89339108 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
mutaFISH-Ce
Catalog No. For Use With (Application)
89-339-108 mutaFISH-Ce
1 options
Catalog No. 89-339-108 Supplier Abnova™ Supplier No. FP0023
Only null left
Add to Cart
Add to Cart
mutaFISH™ EGFRwt RNA Probes is designed to detect human EGFR gene on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.
Specifications
For Use With (Application) mutaFISH-Ce
Includes Content:
1. RT EGFR Primer
2. mutaFISH™ EGFRwt RNA Probe
3. Detection Probe-FITC