Learn More
Abnova™ mutaFISH™ ERCC1wt RNA Probes
Catalog No. 89339109 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
mutaFISH-Fr
Catalog No. For Use With (Application)
89-339-109 mutaFISH-Fr
1 options
Catalog No. 89-339-109 Supplier Abnova™ Supplier No. FP0024
Add to Cart
Add to Cart
mutaFISH™ ERCC1wt RNA Probes is designed to detect human ERCC1 gene on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.
Specifications
For Use With (Application) mutaFISH-Fr
Includes Content:
1. RT ERCC1 Primer
2. mutaFISH™ ERCC1wt RNA Probe
3. Detection Probe-Texas Red X