Learn More
Abnova™ mutaFISH™ HER2wt RNA Probes
Catalog No. 89339116 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
mutaFISH-P
Catalog No. For Use With (Application)
89-339-116 mutaFISH-P
1 options
Catalog No. 89-339-116 Supplier Abnova™ Supplier No. FP0022
Add to Cart
Add to Cart
mutaFISH™ HER2wt RNA Probes is designed to detect human HER2 gene on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.
Specifications
For Use With (Application) mutaFISH-P
Includes Content:
1. RT HER2 Primer
2. mutaFISH™ HER2wt RNA Probe
3. Detection Probe-Texas Red X