Learn More
Abnova™ mutaFISH™ HLA-ABCwt RNA Probes
Catalog No. 89339110 Shop All Abnova Corporation Products
Change view
Click to view available options
For Use With (Application):
mutaFISH-Ce
Catalog No. For Use With (Application)
89-339-110 mutaFISH-Ce
1 options
Catalog No. 89-339-110 Supplier Abnova™ Supplier No. FP0021
Add to Cart
Add to Cart
mutaFISH™ HLA-ABCwt RNA Probes is designed to detect human HLA-A, B, and C gene on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.
Specifications
For Use With (Application) mutaFISH-Ce
Includes Content:
1. RT HLA-ABC Primer
2. mutaFISH™ HLA-ABCwt RNA Probe
3. Detection Probe-Aqua 431