Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ mutaFISH™ IDH1 R132H R132wt IDH2 R172K R172wt RNA Probes

Catalog No. 89333455 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Probe Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-455 1 Probe Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-455 Supplier Abnova™ Supplier No. FP0012
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

mutaFISH™ IDH1 R132H R132wt IDH2 R172K R172wt RNA Probes is designed to detect human IDH1 R132H gene mutation and IDH2 R172K gene mutation on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.

Specifications

Content And Storage Store at -20°C.
Description isocitrate dehydrogenase 1 (NADP+), soluble
Label or Dye Cy3, Cy5, Texas Red
For Use With (Application) mutaFISH-Ce
Location IDH1: Exon 4 (R132), IDH2: Exon 4 (R172)
Product Type DNA/RNA
Quantity 1 Probe Set
Size 1 Probe Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.