Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ mutaFISH™ PSCAwt RNA Probes

Catalog No. 89339167 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Probe Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-339-167 1 Probe Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-339-167 Supplier Abnova™ Supplier No. FP0032
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

mutaFISH™ PSCAwt RNA Probes is designed to detect human PSCA gene on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.

Specifications

Content And Storage Store at -20°C.
Aliquot to avoid repeated freezing and thawing.
For Use With (Application) mutaFISH-Ce
Gene ID (Entrez) 8000
Includes Content:

1. RT PSCA Primer

2. mutaFISH™ PSCAwt RNA Probe

3. Detection Probe-FITC
Quantity 1 Probe Set
Target Human
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.