Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Abnova™ mutaFISH™ TERTwt RNA Probes

Catalog No. 89334095 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
1 Probe Set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-334-095 1 Probe Set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-334-095 Supplier Abnova™ Supplier No. FP0016
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Labeled FISH probes for identification of gene amplification using Fluoresecent In Situ Hybridization Technique.

mutaFISH™ TERTwt RNA Probes is designed to detect human TERT wildtype gene on single strand RNA in cells using padlock probe and in situ rolling-circle amplification technology.

Specifications

Content And Storage Store at -20°C.
Description telomerase reverse transcriptase
Label or Dye Texas Red
For Use With (Application) mutaFISH-Ce
Location 5p15.33
Product Type DNA/RNA
Quantity 1 Probe Set
Size 1 Probe Set
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.