Learn More
Sigma Aldrich Fine Chemicals Biosciences Mutanolysin from Streptomyces globisporus ATCC 21553 free of DNA contaminants, suitable for Microbiome research, lyophilized powder, >=4000 units/mg protein (biuret) | 55466-22-3 | MFCD00131635 | 10KU
Catalog No. NC1571641
Encompass
Mutanolysin from Streptomyces globisporus ATCC 21553 free of DNA contaminants, suitable for Microbiome research, lyophilized powder, >=4000 units/mg protein (biuret) | 55466-22-3 | MFCD00131635 | 10KU
Catalog No. NC1571641 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. SAE009210KU
Mutanolysin from Streptomyces globisporus ATCC 21553 free of DNA contaminants, suitable for Microbiome research, lyophilized powder, >=4000 units/mg protein (biuret) | 55466-22-3 | MFCD00131635 | 10KU