Learn More
Medchemexpress LLC Mutant EGFR inhibitor | 1421373-62-7 | 99.4% | 520.03 | 50 MG
SDP
Catalog No. 5000347847
Encompass_Preferred
Mutant EGFR inhibitor (Example 25) is a potent mutant EGFR inhibitor. It inhibits FGFRWT with an IC50 of 0.41 nM. This inhibitor also targets EGFRL858R, EGFRExon 19 deletion, and EGFRT790M. It is intended for the study of diseases mediated by EGFR, especially cancers, and is for research use only, not for sale to patients.
  • Potent mutant EGFR inhibitor
  • Inhibits FGFRWT with an IC50 of 0.41 nM
  • Targets EGFRL858R, EGFRExon 19 deletion, and EGFRT790M
  • Used for the study of diseases mediated by EGFR, particularly cancers
  • For research use only
Catalog No. 50-003-47847 Supplier Medchemexpress LLC Supplier No. HY1398450MG
Mutant EGFR inhibitor (Example 25) is a potent mutant EGFR inhibitor. It inhibits FGFRWT with an IC50 of 0.41 nM. This inhibitor also targets EGFRL858R, EGFRExon 19 deletion, and EGFRT790M. It is intended for the study of diseases mediated by EGFR, especially cancers, and is for research use only, not for sale to patients.
  • Potent mutant EGFR inhibitor
  • Inhibits FGFRWT with an IC50 of 0.41 nM
  • Targets EGFRL858R, EGFRExon 19 deletion, and EGFRT790M
  • Used for the study of diseases mediated by EGFR, particularly cancers
  • For research use only