Learn More
Medchemexpress LLC HY-13984 5mg Medchemexpress, Mutant EGFR inhibitor CAS:1421373-62-7 Purity:>98%
SDP
Catalog No. 502081541
Encompass_Preferred
Medchemexpress, HY-13984 5mg Mutant EGFR inhibitor CAS:1421373-62-7 Mutant EGFR inhibitor is a potent and selective mutant EGFR inhibitor extracted from patent WO 2013014448 A1; inhibits EGFRL858R, EGFRExon 19 deletion and EGFRT790M. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1541 Supplier Medchemexpress LLC Supplier No. HY139845MG
Medchemexpress, HY-13984 5mg Mutant EGFR inhibitor CAS:1421373-62-7 Mutant EGFR inhibitor is a potent and selective mutant EGFR inhibitor extracted from patent WO 2013014448 A1; inhibits EGFRL858R, EGFRExon 19 deletion and EGFRT790M. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.