Learn More
TARGETMOL CHEMICALS INC MUTANT IDH1-IN-2 5MG
SDP
Catalog No. 502581481
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mutant IDH1-IN-2 is a inhibitor of mutant Isocitrate dehydrogenase (IDH) proteins with IC50 of 16.6 nM in Fluorescence biochemical assay IC50 of <22 nM in LS-MS biochemical assay. purity: 99%
Catalog No. 50-258-1481 Supplier TARGETMOL CHEMICALS INC Supplier No. T121285MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mutant IDH1-IN-2 is a inhibitor of mutant Isocitrate dehydrogenase (IDH) proteins with IC50 of 16.6 nM in Fluorescence biochemical assay IC50 of <22 nM in LS-MS biochemical assay. purity: 99%