Learn More
Medchemexpress LLC Mutant IDH1 inhibitor | 1429180-08-4 | 99.1% | 466.58 | 10 MM 1 ML
SDP
Catalog No. 5000347844
Encompass_Preferred
Mutant IDH1 inhibitor is a potent mutant IDH1 R132H inhibitor with an IC50 of < 72 nM. It is used for the treatment of diseases or disorders associated with such mutant IDH proteins, including cell-proliferation disorders, such as cancer.
  • Potent mutant IDH1 R132H inhibitor
  • IC50 of < 72 nM
  • Treats diseases associated with mutant IDH proteins
  • Applicable for cell-proliferation disorders including cancer
Catalog No. 50-003-47844 Supplier Medchemexpress LLC Supplier No. HY1397210MM/1ML
Mutant IDH1 inhibitor is a potent mutant IDH1 R132H inhibitor with an IC50 of < 72 nM. It is used for the treatment of diseases or disorders associated with such mutant IDH proteins, including cell-proliferation disorders, such as cancer.
  • Potent mutant IDH1 R132H inhibitor
  • IC50 of < 72 nM
  • Treats diseases associated with mutant IDH proteins
  • Applicable for cell-proliferation disorders including cancer