Learn More
Medchemexpress LLC Mutant IDH1 inhibitor | 1429180-08-4 | 99.1% | 466.58 | 50 MG
SDP
Catalog No. 5000349876
Encompass_Preferred
Mutant IDH1 inhibitor is a potent mutant IDH1 R132H inhibitor with an IC50 of < 72 nM. It is used for research purposes in the treatment of diseases or disorders associated with mutant IDH proteins, including cell-proliferation disorders like cancer.
  • Potent mutant IDH1 R132H inhibitor
  • IC50 of < 72 nM
  • Used for research purposes
  • Treats diseases associated with mutant IDH proteins
  • Including cell-proliferation disorders like cancer
Catalog No. 50-003-49876 Supplier Medchemexpress LLC Supplier No. HY1397250MG
Mutant IDH1 inhibitor is a potent mutant IDH1 R132H inhibitor with an IC50 of < 72 nM. It is used for research purposes in the treatment of diseases or disorders associated with mutant IDH proteins, including cell-proliferation disorders like cancer.
  • Potent mutant IDH1 R132H inhibitor
  • IC50 of < 72 nM
  • Used for research purposes
  • Treats diseases associated with mutant IDH proteins
  • Including cell-proliferation disorders like cancer