Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mutant IDH1 inhibitor | 1429180-08-4 | 99.4% | C25H34N6O3 | 10MG
SDP

Catalog No. 5000262022
Encompass_Preferred

Mutant IDH1 inhibitor is a research-grade small molecule that selectively inhibits mutant isocitrate dehydrogenase 1 (IDH1) R132H with sub-100 nM potency. It is supplied as a high-purity solid for biochemical assays and in vitro studies, and is accompanied by documentation for quality control and safe handling.

  • High potency against mutant IDH1 R132H (IC50 < 72 nM).
  • High purity suitable for research (99.4% by HPLC).
  • Molecular formula C25H34N6O3; molecular weight 466.58 g/mol.
  • Supplied as a solid, white to light yellow, in small laboratory packaging.
  • Recommended storage: powder long-term at -20°C, short-term at 4°C.
  • Includes downloadable certificate of analysis and safety data sheet for traceability and safety.

Catalog No. 50-002-62022 Supplier Medchemexpress LLC Supplier No. HY1397210MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mutant IDH1 inhibitor is a research-grade small molecule that selectively inhibits mutant isocitrate dehydrogenase 1 (IDH1) R132H with sub-100 nM potency. It is supplied as a high-purity solid for biochemical assays and in vitro studies, and is accompanied by documentation for quality control and safe handling.

  • High potency against mutant IDH1 R132H (IC50 < 72 nM).
  • High purity suitable for research (99.4% by HPLC).
  • Molecular formula C25H34N6O3; molecular weight 466.58 g/mol.
  • Supplied as a solid, white to light yellow, in small laboratory packaging.
  • Recommended storage: powder long-term at -20°C, short-term at 4°C.
  • Includes downloadable certificate of analysis and safety data sheet for traceability and safety.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.