Learn More
Medchemexpress LLC HY-13972 5mg Medchemexpress, Mutant IDH1 inhibitor CAS:1429180-08-4 Purity:>98%
SDP
Catalog No. 502081539
Encompass_Preferred
Medchemexpress, HY-13972 5mg Mutant IDH1 inhibitor CAS:1429180-08-4 Mutant IDH1 inhibitor is a potent mutant IDH1 R132H inhibitor with IC50 of < 72 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-1539 Supplier Medchemexpress LLC Supplier No. HY139725MG
Medchemexpress, HY-13972 5mg Mutant IDH1 inhibitor CAS:1429180-08-4 Mutant IDH1 inhibitor is a potent mutant IDH1 R132H inhibitor with IC50 of < 72 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.