Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mutant p53 modulator-1 | 2746371-35-5 | 99.89% | C27H32F4N8O2 | 25 MG
SDP

Catalog No. 5000356485
Encompass_Preferred

Mutant p53 modulator-1 is a compound that acts as a mutant p53 modulator. It reduces the progression of cancers containing a p53 mutation. This product is for research use only and not intended for patient use.

  • Reduces progression of cancers with p53 mutation
  • For research use only
  • Powder storage at -20°C for 3 years, or 4°C for 2 years
  • In solvent storage at -80°C for 6 months, or -20°C for 1 month
  • Appears as a light yellow to brown solid
  • Purity of 99.89% by HPLC

Catalog No. 50-003-56485 Supplier Medchemexpress LLC Supplier No. 25MGHY145759
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mutant p53 modulator-1 is a compound that acts as a mutant p53 modulator. It reduces the progression of cancers containing a p53 mutation. This product is for research use only and not intended for patient use.

  • Reduces progression of cancers with p53 mutation
  • For research use only
  • Powder storage at -20°C for 3 years, or 4°C for 2 years
  • In solvent storage at -80°C for 6 months, or -20°C for 1 month
  • Appears as a light yellow to brown solid
  • Purity of 99.89% by HPLC

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.