Learn More
Aviva Systems Biology Corp MUTED Antibody - C-terminal region (ARP63301_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP63301_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MUTED. It was validated on Western Blot. . Alternate names: DKFZp686E2287, MU, MUTEDApplicationWB. Species reactivity: Cow, Dog, Guinea Pig, Horse, Human, Mouse, Rabbit, Rat
Catalog No. 50-287-934