Learn More
Cayman Chemical Mutnt IDH1-IN-1 50mg
SDP
Catalog No. 502979671
Encompass_Preferred
An inhibitor of mutant IDH1 (IC50s = 42, 4, and 80 nM for IDH1R132H/R132H, IDH1R132C/R132C, and IDH1R132H/WT, respectively, for conversion of α-KG to 2-HG); selective for IDH1 mutant conversion of α-KG to 2-HT over IDH1WT/WT conversion of isocitrate to α-KG (IC50 = 1,998 nM) and over IDH2 (IC50s = >10,000 nM); inhibits 2-HG production in vitro (IC50 = 81.5 nM)
Catalog No. 50-297-9671 Supplier Cayman Chemical Supplier No. 1839450 MG
An inhibitor of mutant IDH1 (IC50s = 42, 4, and 80 nM for IDH1R132H/R132H, IDH1R132C/R132C, and IDH1R132H/WT, respectively, for conversion of α-KG to 2-HG); selective for IDH1 mutant conversion of α-KG to 2-HT over IDH1WT/WT conversion of isocitrate to α-KG (IC50 = 1,998 nM) and over IDH2 (IC50s = >10,000 nM); inhibits 2-HG production in vitro (IC50 = 81.5 nM)