Learn More
AdipoGen Mutolide
SDP
Supplier:  AdipoGen BVT0070M005
Encompass_Preferred
Chemical. CAS 277749-34-5. Formula C14H20O4. MW 252.3. Isolated from Sphaeropsidales sp. Antibiotic. Antibacterial. Phytotoxic.
Catalog No. 50-256-2118