Learn More
Medchemexpress LLC Muzolimine | 55294-15-0 | 98.2% | 272.13 g/mol | C11H11Cl2N3O | 50 MG
SDP
Supplier:  Medchemexpress LLC HY10661650MG
Encompass_Preferred
Muzolimine is a research-grade loop diuretic supplied as a light yellow to yellow solid for laboratory and analytical applications. The material has a reported purity of 98.22% and a molecular weight of 272.13 g/mol. Multiple solid pack sizes and a DMSO solution format are available, with storage recommendations for both powder and solvent forms.
Catalog No. 50-002-41094