Learn More
Medchemexpress LLC Muzolimine | 55294-15-0 | MFCD00867325 | 98.2% | 272.13 g/mol | C11H11Cl2N3O | 1 ML

Supplier: Medchemexpress LLC HY10661610MM 1ML
Muzolimine (CAS 55294-15-0) is a pyrazolone diuretic supplied as a ready-to-use 10 mM solution in DMSO for research applications. The product is offered at high analytical purity and includes molecular data for experimental use.
- 10 mM solution in DMSO, 1 mL vial.
- Purity 98.2% as stated on product page.
- Molecular formula C11H11Cl2N3O; molecular weight 272.13 g/mol.
- Provided ready for dilution or direct use in assays.
- Suitable for pharmacological and diuresis research applications.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.