Learn More
Med Vet International Jorvet Buster Nylon Basket Muzzle for Dogs, Size #4, J0884C
Catalog No. 5000495137
Encompass_Preferred
The Jorvet Buster Nylon basket muzzle was designed to prevent bites from dogs. This dog muzzle is made of a strong nylon plastic with a reinforced nose and leather straps. This muzzle is a size #4.
Catalog No. 50-004-95137 Supplier Med Vet International Supplier No. JOR884C
The Jorvet Buster Nylon basket muzzle was designed to prevent bites from dogs. This dog muzzle is made of a strong nylon plastic with a reinforced nose and leather straps. This muzzle is a size #4.