Learn More
Med Vet International Jorvet Nylon Basket Muzzle for Dogs, Size #7, J0884F
Catalog No. 5000495216
Encompass_Preferred
The Jorvet Buster Nylon basket muzzle was designed to prevent bites from dogs. This dog muzzle is made of a strong nylon plastic with a reinforced nose and leather straps. This muzzle is a size #7.
Catalog No. 50-004-95216 Supplier Med Vet International Supplier No. JOR884F
The Jorvet Buster Nylon basket muzzle was designed to prevent bites from dogs. This dog muzzle is made of a strong nylon plastic with a reinforced nose and leather straps. This muzzle is a size #7.