Learn More
ATCC Mv 1 Lu (NBL-7) Mink Lung Epithelial Cell Line (Neovison vison) - ATCC CCL-64
Supplier:  ATCC CCL64
Encompass
Mv 1 Lu (NBL-7) is an adherent mink lung epithelial cell line established in May 1964 from nearly full-term Aleutian mink fetuses (Neovison vison). It is commonly used as a transfection host and for focus-forming assays with murine and feline sarcoma viruses, and is suitable for 3D cell culture applications.
  • Established from lung tissue of Aleutian mink fetuses.
  • Epithelial morphology; adherent growth.
  • Suitable for focus-forming assays and transfection studies.
  • Culture medium: Eagle's Minimum Essential Medium (EMEM) supplemented with 10% fetal bovine serum.
  • Incubation: 37°C, 5% CO2.
  • Karyotype: majority of cells near diploid; some cells show a dicentric chromosome.
Catalog No. 50-238-3580