Learn More
Molecular Devices Molecular Devices SpectraDrop Micro-Volume Refills, 24-Well, Teflon-Coated, Pack of 3
Catalog No. 5000014366
Encompass_Preferred
Molecular Devices SpectraDrop Micro-Volume 24-well refill kit includes three low-volume, Teflon-coated bottom slides to replace worn slides in SpectraDrop micro-volume assemblies. Slides are designed for precise droplet placement and reproducible low-volume absorbance and quantitation using 2 μL or 4 μL configurations.
  • Contains three low-volume 24-well bottom slides.
  • Teflon-coated bottom slides for precise droplet placement.
  • Compatible with micro-volume adapters and cover slides.
  • Supports 2 μL and 4 μL low-volume configurations (system-level).
  • Intended for micro-volume absorbance and sample quantitation.
Catalog No. 50-000-14366 Supplier Molecular Devices Supplier No. 02006263
Molecular Devices SpectraDrop Micro-Volume 24-well refill kit includes three low-volume, Teflon-coated bottom slides to replace worn slides in SpectraDrop micro-volume assemblies. Slides are designed for precise droplet placement and reproducible low-volume absorbance and quantitation using 2 μL or 4 μL configurations.
  • Contains three low-volume 24-well bottom slides.
  • Teflon-coated bottom slides for precise droplet placement.
  • Compatible with micro-volume adapters and cover slides.
  • Supports 2 μL and 4 μL low-volume configurations (system-level).
  • Intended for micro-volume absorbance and sample quantitation.