Learn More
Medchemexpress LLC HY-113534 10mg Medchemexpress, MV1 CAS:1001600-54-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY11353410MG
Encompass_Preferred
Medchemexpress, HY-113534 10mg MV1 CAS:1001600-54-9 MV1 is an antagonist of IAP (inhibitor of apoptosis protein), leads to protein knockdown of HaloTag-fused proteins when combined with HaloTag ligand. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9259