Learn More
Labtech Inc MV5 Automated Concentrator. 54 positions supporting 10 or 15ml concentration vials simultaneously, nitrogen blowing needle can automated up-down moving
Catalog No. 5000496198
Encompass_Preferred
MV5 Automated Concentrator. 54 positions supporting 10 or 15ml concentration vials simultaneously, nitrogen blowing needle can automated up-down moving
Catalog No. 50-004-96198 Supplier Labtech Inc Supplier No. MV54
MV5 Automated Concentrator. 54 positions supporting 10 or 15ml concentration vials simultaneously, nitrogen blowing needle can automated up-down moving