Learn More
Aviva Systems Biology Corp MVD Antibody - N-terminal region (ARP61094_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP61094_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MVD. It was validated on Western Blot. The immunogen is a synthetic peptide directed towards the N terminal region of human MVD. Alternate names: FP17780, MPDApplicationWB, IHC. Species reactivity: Human
Catalog No. 50-287-249