Learn More
Cryo Associates MVE 1539P-190AF-GB FULL AUTO
Catalog No. NC3315913
Encompass
  • Advanced automatic fill liquid nitrogen freezer
  • Designed for cryogenic vapor phase storage
  • Maintains -190 °C top box temperature
  • 39,200 vials capacity (1.2/2.0 ml)
  • 797 L LN2 capacity without inventory
  • 133 L LN2 capacity at vapor platform
  • 20 days static hold time in vapor
  • 118 days static hold time with liquid full
  • 17.5 in neck opening
  • 30.8 in usable internal height
  • 38.5 in inner diameter
  • 63.3 in overall height
  • 39.2 in liftover height
  • 42.0 in door width requirement
  • 720 lb empty weight
  • 2140 lb full weight with liquid
  • TEC 3000 controller or touch-screen option
  • MDD 93/42/EEC, UL/C-UL safety compliance
  • Two years parts warranty
  • Five years vacuum warranty
Catalog No. NC3315913 Supplier Cryo Associates Supplier No. 11894505
Add to Cart
Add to Cart
Request a Quote
  • Advanced automatic fill liquid nitrogen freezer
  • Designed for cryogenic vapor phase storage
  • Maintains -190 °C top box temperature
  • 39,200 vials capacity (1.2/2.0 ml)
  • 797 L LN2 capacity without inventory
  • 133 L LN2 capacity at vapor platform
  • 20 days static hold time in vapor
  • 118 days static hold time with liquid full
  • 17.5 in neck opening
  • 30.8 in usable internal height
  • 38.5 in inner diameter
  • 63.3 in overall height
  • 39.2 in liftover height
  • 42.0 in door width requirement
  • 720 lb empty weight
  • 2140 lb full weight with liquid
  • TEC 3000 controller or touch-screen option
  • MDD 93/42/EEC, UL/C-UL safety compliance
  • Two years parts warranty
  • Five years vacuum warranty