Learn More
Princeton Cryotech MVE HECO 800 SERIES FREEZER
Catalog No. NC2094033
Encompass_Preferred
MVE HEco 800 Series Freezer - Vapor phase storage for 18,200 1.2 & 2.0 mL vials
This item is not returnable. View return policy
Catalog No. NC2094033 Supplier Princeton Cryotech Supplier No. 21093058
Add to Cart
Add to Cart
Request a Quote
MVE HEco 800 Series Freezer - Vapor phase storage for 18,200 1.2 & 2.0 mL vials