Learn More
Aviva Systems Biology Corp MVK Antibody - middle region (ARP56110_P050) | 100uL
SDP
Supplier:  Aviva Systems Biology Corp ARP56110_P050
Encompass_Preferred
This is a rabbit polyclonal antibody against MVK. It was validated on Western Blot using a cell lysate as a positive control.The immunogen is a synthetic peptide directed towards the middle region of human MVK. Alternate names: LRBP, MVLK, MKApplicationWB. Species reactivity: Cow, Dog, Goat, Guinea Pig, Horse, Human, Mouse, Pig, Rabbit, Rat, Yeast, Zebrafish
Catalog No. 50-014-61723