Learn More
Sigma Aldrich Fine Chemicals Biosciences Surface Sampling Device
Catalog No. NC1781716
Encompass
The MVP ICON Surface Sampling Device is a self-contained device for use with the MVP ICON system for quality and HACCP-related monitoring of the environment. The MVP ICON system detects food and microbiological residues by measuring Adenosine triphosphate (ATP) using luciferase bioluminescence. ATP is found in most food residues and all bacteria, yeast and mold cells.
Catalog No. NC1781716 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. 64003100
The MVP ICON Surface Sampling Device is a self-contained device for use with the MVP ICON system for quality and HACCP-related monitoring of the environment. The MVP ICON system detects food and microbiological residues by measuring Adenosine triphosphate (ATP) using luciferase bioluminescence. ATP is found in most food residues and all bacteria, yeast and mold cells.